The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194286.3 | NP_919262.2 | 7 - 8 | 998 | 75 |
GenBank mRNA | AB058756.1 | - | 7 - 8 | 902 | 75 |
AK291361.1 | - | 7 - 8 | 998 | 75 | |
BC152471.1 | - | 7 - 8 | 913 | 75 | |
BC167162.1 | - | 7 - 8 | 902 | 75 | |
BC167163.1 | - | 7 - 8 | 902 | 75 | |
KC995185.1 | - | 7 - 8 | 610 | 75 | |
KC995186.1 | - | 7 - 8 | 610 | 75 |