The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042574.2 | NP_001036039.1 | 8 - 9 | 78 | |
NM_022769.4 | NP_073606.3 | 8 - 9 | 78 | ||
XM_005254968.2 | XP_005255025.1 | 8 - 9 | 78 | ||
XM_011521906.2 | XP_011520208.1 | 7 - 8 | 78 | ||
GenBank mRNA | AK025521.1 | - | 1 - 2 | 78 | |
AK074170.1 | - | 5 - 6 | 78 | ||
AK090443.1 | - | 8 - 9 | 78 | ||
AK225300.1 | - | 1 - 2 | 78 | ||
AY360173.1 | - | 8 - 9 | 78 | ||
BP238198.1 | - | 1 - 2 | 58 | 78 |