The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042574.2 | NP_001036039.1 | 14 - 15 | 1835 | 149 |
GenBank mRNA | AK025521.1 | - | 7 - 8 | 1002 | 149 |
AK074170.1 | - | 11 - 12 | 1306 | 149 | |
AK090443.1 | - | 14 - 15 | 1795 | 149 | |
AK225300.1 | - | 7 - 8 | 1002 | 149 | |
BC074730.2 | - | 5 - 6 | 778 | 149 |