The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006011.3 | NP_006002.1 | 4 - 5 | 699 | 74 |
XM_017022642.1 | XP_016878131.1 | 4 - 5 | 862 | 74 | |
XM_017022643.1 | XP_016878132.1 | 3 - 4 | 635 | 74 | |
GenBank mRNA | AK313924.1 | - | 4 - 5 | 647 | 74 |
BC069584.1 | - | 4 - 5 | 568 | 74 | |
BC096202.3 | - | 4 - 5 | 568 | 74 | |
BC096203.3 | - | 4 - 5 | 568 | 74 | |
BC096204.3 | - | 4 - 5 | 568 | 74 | |
BC096205.3 | - | 3 - 4 | 505 | 74 | |
U33551.1 | - | 4 - 5 | 544 | 74 | |
U82762.1 | - | 4 - 5 | 569 | 74 |