The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003358.2 | NP_003349.1 | 2 - 3 | 685 | 139 |
XM_017015107.1 | XP_016870596.1 | 2 - 3 | 344 | 139 | |
GenBank mRNA | AK314847.1 | - | 2 - 3 | 290 | 139 |
BC038711.1 | - | 2 - 3 | 686 | 139 | |
D50840.1 | - | 2 - 3 | 525 | 139 |