The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002486.4 | NP_002477.1 | 14 - 15 | 1828 | 105 |
XM_017014735.1 | XP_016870224.1 | 14 - 15 | 1666 | 105 | |
XM_017014736.1 | XP_016870225.1 | 14 - 15 | 1379 | 105 | |
XM_017014737.1 | XP_016870226.1 | 14 - 15 | 1367 | 105 | |
XM_017014738.1 | XP_016870227.1 | 13 - 14 | 1367 | 105 | |
GenBank mRNA | AB209233.1 | - | 14 - 15 | 1408 | 105 |
AK312807.1 | - | 14 - 15 | 1828 | 105 | |
BC001450.2 | - | 14 - 15 | 1432 | 105 | |
D32002.1 | - | 14 - 15 | 1633 | 105 | |
X80030.1 | - | 14 - 15 | 1399 | 105 |