The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145862.1 | NP_001139334.1 | 14 - 15 | 1713 | 78 |
NM_181873.3 | NP_870988.2 | 13 - 14 | 1455 | 78 | |
XM_006711137.1 | XP_006711200.1 | 13 - 14 | 1432 | 78 | |
GenBank mRNA | AK097000.1 | - | 13 - 14 | 1432 | 78 |
AK298775.1 | - | 14 - 15 | 1682 | 78 | |
BC032655.1 | - | 7 - 8 | 787 | 78 | |
BC064372.1 | - | 12 - 13 | 1674 | 78 | |
BC100818.2 | - | 13 - 14 | 1373 | 78 | |
BC100819.2 | - | 13 - 14 | 1373 | 78 | |
BC100820.1 | - | 13 - 14 | 1373 | 78 | |
BC100821.2 | - | 13 - 14 | 1326 | 78 |