The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145862.1 | NP_001139334.1 | 5 - 6 | 712 | 83 |
NM_181873.3 | NP_870988.2 | 4 - 5 | 454 | 83 | |
XM_006711137.1 | XP_006711200.1 | 4 - 5 | 431 | 83 | |
XM_011509099.2 | XP_011507401.1 | 5 - 6 | 605 | 83 | |
GenBank mRNA | AK097000.1 | - | 4 - 5 | 431 | 83 |
AK296094.1 | - | 4 - 5 | 551 | 83 | |
AK304691.1 | - | 4 - 5 | 528 | 83 | |
AK304820.1 | - | 5 - 6 | 682 | 83 | |
BC032655.1 | - | 4 - 5 | 393 | 83 | |
BC064372.1 | - | 5 - 6 | 684 | 83 | |
BC094756.1 | - | 5 - 6 | 693 | 83 | |
BC100818.2 | - | 4 - 5 | 372 | 83 | |
BC100819.2 | - | 4 - 5 | 372 | 83 | |
BC100820.1 | - | 4 - 5 | 372 | 83 | |
BC100821.2 | - | 4 - 5 | 372 | 83 | |
U78556.1 | - | 5 - 6 | 647 | 83 | |
U78557.1 | - | 5 - 6 | 647 | 83 |