The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017895.7 | NP_060365.7 | 10 - 11 | 1289 | 65 |
XM_011528888.2 | XP_011527190.1 | 11 - 12 | 1353 | 65 | |
GenBank mRNA | AF193054.1 | - | 7 - 8 | 847 | 65 |
AF336851.1 | - | 4 - 5 | 523 | 65 | |
AK000603.1 | - | 1 - 2 | 64 | 65 | |
AK022979.1 | - | 10 - 11 | 1159 | 65 | |
AK025891.1 | - | 6 - 7 | 654 | 65 | |
AK223116.1 | - | 1 - 2 | 64 | 65 | |
AK300135.1 | - | 10 - 11 | 1168 | 65 | |
AL832131.1 | - | 10 - 11 | 1205 | 65 | |
AY044431.1 | - | 10 - 11 | 1228 | 65 | |
BC009304.1 | - | 10 - 11 | 1147 | 65 | |
BC011927.2 | - | 10 - 11 | 1143 | 65 | |
BC016060.1 | - | 10 - 11 | 1143 | 65 | |
BC126287.1 | - | 10 - 11 | 1278 | 65 | |
BC130275.1 | - | 10 - 11 | 1278 | 65 | |
BC144125.1 | - | 10 - 11 | 1278 | 65 | |
EU718675.1 | - | 10 - 11 | 1196 | 65 |