Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DDX27
- Entrez Gene ID
-
55661
- Gene Name
- DEAD-box helicase 27
- Gene Aliases
-
DRS1, Drs1p, HSPC259, PP3241, RHLP, dJ686N3.1
- Chromosome Location
-
Chr.20: 49219295 - 49244077 on Build GRCh38
- Gene Symbol
- SS18
- Entrez Gene ID
-
6760
- Gene Name
- SS18, nBAF chromatin remodeling complex subunit
- Gene Aliases
-
SSXT, SYT
- Chromosome Location
-
Chr.18: 26016253 - 26091217 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF161377.1 |
- |
2 - 3 |
312 |
113 |
View Full Product Details