The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000449.3 | NP_000440.1 | 3 - 4 | 341 | 76 |
NM_001025603.1 | NP_001020774.1 | 3 - 4 | 334 | 76 | |
XM_005245405.1 | XP_005245462.1 | 3 - 4 | 396 | 76 | |
XM_005245406.3 | XP_005245463.1 | 3 - 4 | 353 | 76 | |
XM_011509847.1 | XP_011508149.1 | 3 - 4 | 400 | 76 | |
XM_011509848.1 | XP_011508150.1 | 3 - 4 | 403 | 76 | |
XM_011509849.1 | XP_011508151.1 | 3 - 4 | 336 | 76 | |
XM_011509850.1 | XP_011508152.1 | 3 - 4 | 463 | 76 | |
XM_017001999.1 | XP_016857488.1 | 3 - 4 | 396 | 76 | |
XM_017002000.1 | XP_016857489.1 | 3 - 4 | 427 | 76 | |
GenBank mRNA | AK223526.1 | - | 1 - 2 | 133 | 76 |
AK302891.1 | - | 3 - 4 | 334 | 76 | |
AK309072.1 | - | 3 - 4 | 353 | 76 | |
AK312289.1 | - | 3 - 4 | 76 | ||
BC017471.1 | - | 3 - 4 | 268 | 76 | |
BX423384.2 | - | 3 - 4 | 367 | 76 | |
X85786.1 | - | 2 - 3 | 281 | 76 |