Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIZ
Entrez Gene ID
55857
Gene Name
kizuna centrosomal protein
Gene Aliases
C20orf19, HT013, Kizuna, NCRNA00153, PLK1S1, RP69
Chromosome Location
Chr.20: 21125980 - 21246628 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163022.1 NP_001156494.1 10 - 11 1849 154
NM_001163023.1 NP_001156495.1 9 - 10 1759 154
NM_001276389.1 NP_001263318.1 9 - 10 1786 154
NM_018474.4 NP_060944.3 11 - 12 2012 154
XM_011529298.2 XP_011527600.1 11 - 12 1943 154
XM_017027953.1 XP_016883442.1 12 - 13 2030 154
XM_017027954.1 XP_016883443.1 10 - 11 1790 154
GenBank mRNA AF220187.1 - 7 - 8 1044 154
AK025476.1 - 11 - 12 1943 154
AK293166.1 - 9 - 10 1690 154
AK293567.1 - 9 - 10 1730 154
AK295355.1 - 10 - 11 1749 154
AL110196.1 - 7 - 8 1237 154
BC045826.1 - 10 - 11 1736 154
BC065550.1 - 9 - 10 1593 154
BC090879.2 - 7 - 8 1417 154
BC105093.1 - 11 - 12 1915 154
BC113370.1 - 11 - 12 1915 154

View Full Product Details