Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF20
Entrez Gene ID
51230
Gene Name
PHD finger protein 20
Gene Aliases
C20orf104, GLEA2, HCA58, NZF, TDRD20A, TZP
Chromosome Location
Chr.20: 35771999 - 35950377 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016436.4 NP_057520.2 12 - 13 1937 92
XM_005260421.2 XP_005260478.1 9 - 10 1547 92
XM_011528843.1 XP_011527145.1 11 - 12 1538 92
XM_011528844.1 XP_011527146.1 9 - 10 1609 92
XM_017027864.1 XP_016883353.1 13 - 14 2019 92
XM_017027865.1 XP_016883354.1 13 - 14 2220 92
XM_017027866.1 XP_016883355.1 12 - 13 2140 92
XM_017027867.1 XP_016883356.1 12 - 13 1975 92
XM_017027868.1 XP_016883357.1 10 - 11 1761 92
XM_017027869.1 XP_016883358.1 12 - 13 1934 92
XM_017027870.1 XP_016883359.1 12 - 13 1940 92
XM_017027871.1 XP_016883360.1 13 - 14 2020 92
GenBank mRNA AF348207.1 - 11 - 12 1862 92
AK097498.1 - 1 - 2 140 92
AK314503.1 - 12 - 13 1901 92
AY027523.1 - 11 - 12 1862 92
BC150178.1 - 12 - 13 1903 92

View Full Product Details