Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF20
Entrez Gene ID
51230
Gene Name
PHD finger protein 20
Gene Aliases
C20orf104, GLEA2, HCA58, NZF, TDRD20A, TZP
Chromosome Location
Chr.20: 35771999 - 35950377 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016436.4 NP_057520.2 2 - 3 197 90
XM_011528843.1 XP_011527145.1 2 - 3 186 90
XM_017027864.1 XP_016883353.1 3 - 4 279 90
XM_017027865.1 XP_016883354.1 3 - 4 480 90
XM_017027866.1 XP_016883355.1 2 - 3 400 90
XM_017027867.1 XP_016883356.1 2 - 3 235 90
XM_017027868.1 XP_016883357.1 2 - 3 186 90
XM_017027869.1 XP_016883358.1 3 - 4 274 90
GenBank mRNA AF220416.1 - 2 - 3 161 90
AF258787.1 - 2 - 3 183 90
AF348207.1 - 1 - 2 122 90
AK054597.1 - 3 - 4 454 90
AK314503.1 - 2 - 3 161 90
AL558423.3 - 1 - 2 120 90
AY027523.1 - 1 - 2 122 90
AY134747.1 - 2 - 3 174 90
BC006415.1 - 2 - 3 137 90
BC016948.1 - 2 - 3 150 90
BC048210.1 - 3 - 4 230 90
BC080598.1 - 2 - 3 161 90
BC093405.1 - 2 - 3 197 90
BC150178.1 - 2 - 3 163 90
BM906582.1 - 2 - 3 186 90
BX643875.1 - 2 - 3 155 90
CD690482.1 - 2 - 3 199 90

View Full Product Details