Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FERMT1
Entrez Gene ID
55612
Gene Name
fermitin family member 1
Gene Aliases
C20orf42, DTGCU2, KIND1, UNC112A, URP1
Chromosome Location
Chr.20: 6074845 - 6123544 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017671.4 NP_060141.3 10 - 11 2054 75
GenBank mRNA AB105105.1 - 10 - 11 2054 75
AF443278.1 - 10 - 11 1623 75
AK092195.1 - 8 - 9 1112 75
AK314394.1 - 10 - 11 1444 75
AY137240.1 - 9 - 10 1265 75
BC035882.1 - 10 - 11 1737 75
BC040545.1 - 9 - 10 1309 75
BX647465.1 - 5 - 6 461 75

View Full Product Details