The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017671.4 | NP_060141.3 | 6 - 7 | 1641 | 130 |
GenBank mRNA | AB105105.1 | - | 6 - 7 | 1641 | 130 |
AF443278.1 | - | 6 - 7 | 1210 | 130 | |
AK092195.1 | - | 4 - 5 | 699 | 130 | |
AY137240.1 | - | 5 - 6 | 852 | 130 | |
BC035882.1 | - | 6 - 7 | 1324 | 130 |