The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079874.1 | NP_001073343.1 | 2 - 3 | 167 | 68 |
NM_006113.4 | NP_006104.4 | 19 - 20 | 1828 | 68 | |
XM_005270360.2 | XP_005270417.1 | 18 - 19 | 1640 | 68 | |
XM_017000054.1 | XP_016855543.1 | 19 - 20 | 1821 | 68 | |
XM_017000055.1 | XP_016855544.1 | 18 - 19 | 1498 | 68 | |
XM_017000056.1 | XP_016855545.1 | 8 - 9 | 704 | 68 | |
GenBank mRNA | AF067817.1 | - | 19 - 20 | 1811 | 68 |
AF118886.1 | - | 18 - 19 | 1632 | 68 | |
AF118887.1 | - | 19 - 20 | 1820 | 68 | |
AK304088.1 | - | 2 - 3 | 167 | 68 | |
AK316295.1 | - | 2 - 3 | 167 | 68 | |
BC143969.1 | - | 19 - 20 | 1911 | 68 |