Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VAV3
Entrez Gene ID
10451
Gene Name
vav guanine nucleotide exchange factor 3
Gene Aliases
-
Chromosome Location
Chr.1: 107571160 - 107964923 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079874.1 NP_001073343.1 8 - 9 740 85
NM_006113.4 NP_006104.4 25 - 26 2401 85
XM_005270360.2 XP_005270417.1 24 - 25 2213 85
XM_017000053.1 XP_016855542.1 23 - 24 2322 85
XM_017000055.1 XP_016855544.1 24 - 25 2071 85
XM_017000056.1 XP_016855545.1 14 - 15 1277 85
GenBank mRNA AF067817.1 - 25 - 26 2384 85
AF118886.1 - 24 - 25 2205 85
AF118887.1 - 25 - 26 2393 85
AK300474.1 - 7 - 8 695 85
AK304088.1 - 8 - 9 740 85
AK316295.1 - 8 - 9 740 85

View Full Product Details