The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018268.3 | NP_060738.2 | 11 - 12 | 1409 | 108 |
XM_005248551.4 | XP_005248608.1 | 9 - 10 | 1202 | 108 | |
XM_005248552.3 | XP_005248609.1 | 11 - 12 | 1376 | 109 | |
XM_011543505.2 | XP_011541807.1 | 11 - 12 | 1171 | 108 | |
XM_017009606.1 | XP_016865095.1 | 11 - 12 | 1819 | 108 | |
GenBank mRNA | AF115511.1 | - | 11 - 12 | 1157 | 108 |
AK001766.1 | - | 11 - 12 | 1355 | 108 | |
AK023977.1 | - | 11 - 12 | 1111 | 108 | |
AK293316.1 | - | 9 - 10 | 894 | 108 | |
AK300058.1 | - | 9 - 10 | 1020 | 108 | |
AK304329.1 | - | 11 - 12 | 1101 | 108 | |
AL834138.1 | - | 5 - 6 | 2090 | 108 | |
BC040241.1 | - | 11 - 12 | 1161 | 108 | |
KU178690.1 | - | 11 - 12 | 1089 | 108 | |
KU178691.1 | - | 11 - 12 | 1104 | 108 |