The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018268.3 | NP_060738.2 | 9 - 10 | 1208 | 148 |
XM_005248551.4 | XP_005248608.1 | 7 - 8 | 1001 | 148 | |
XM_005248552.3 | XP_005248609.1 | 9 - 10 | 1175 | 148 | |
XM_011543505.2 | XP_011541807.1 | 9 - 10 | 970 | 148 | |
XM_011543507.2 | XP_011541809.1 | 9 - 10 | 1175 | 148 | |
XM_017009606.1 | XP_016865095.1 | 9 - 10 | 1618 | 148 | |
GenBank mRNA | AF115511.1 | - | 9 - 10 | 956 | 148 |
AK001766.1 | - | 9 - 10 | 1154 | 148 | |
AK021855.1 | - | 9 - 10 | 994 | 148 | |
AK023977.1 | - | 9 - 10 | 910 | 148 | |
AK293316.1 | - | 7 - 8 | 693 | 148 | |
AK295249.1 | - | 8 - 9 | 815 | 148 | |
AK300058.1 | - | 7 - 8 | 819 | 148 | |
AK304329.1 | - | 9 - 10 | 900 | 148 | |
AL834138.1 | - | 3 - 4 | 1889 | 148 | |
BC040241.1 | - | 9 - 10 | 960 | 148 | |
KU178690.1 | - | 9 - 10 | 888 | 148 | |
KU178691.1 | - | 9 - 10 | 903 | 148 |