The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001129727.2 | NP_001123199.1 | 17 - 18 | 3091 | 61 |
NM_001129728.1 | NP_001123200.1 | 16 - 17 | 3051 | 61 | |
NM_001129729.2 | NP_001123201.1 | 16 - 17 | 3024 | 61 | |
NM_001129731.2 | NP_001123203.1 | 14 - 15 | 3030 | 61 | |
XM_011522985.2 | XP_011521287.1 | 16 - 17 | 3021 | 61 | |
XM_011522986.2 | XP_011521288.1 | 16 - 17 | 3153 | 61 | |
XM_011522987.2 | XP_011521289.1 | 17 - 18 | 3097 | 61 | |
XM_011522988.2 | XP_011521290.1 | 16 - 17 | 2970 | 61 | |
GenBank mRNA | AB197663.1 | - | 15 - 16 | 5287 | 61 |
AB197664.1 | - | 14 - 15 | 5454 | 61 | |
AK024475.1 | - | 16 - 17 | 2986 | 61 | |
AK304723.1 | - | 14 - 15 | 2442 | 61 | |
AL117435.1 | - | 11 - 12 | 1923 | 61 | |
BC054486.1 | - | 14 - 15 | 2965 | 61 | |
BC063501.1 | - | 16 - 17 | 2962 | 61 | |
BC082974.1 | - | 17 - 18 | 3059 | 61 |