The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001129727.2 | NP_001123199.1 | 7 - 8 | 1228 | 60 |
NM_001129728.1 | NP_001123200.1 | 6 - 7 | 1188 | 60 | |
NM_001129729.2 | NP_001123201.1 | 6 - 7 | 1161 | 60 | |
NM_001129731.2 | NP_001123203.1 | 4 - 5 | 1167 | 60 | |
XM_011522985.2 | XP_011521287.1 | 6 - 7 | 1158 | 60 | |
XM_011522986.2 | XP_011521288.1 | 6 - 7 | 1290 | 60 | |
XM_011522987.2 | XP_011521289.1 | 7 - 8 | 1234 | 60 | |
XM_011522988.2 | XP_011521290.1 | 6 - 7 | 1107 | 60 | |
GenBank mRNA | AB197663.1 | - | 5 - 6 | 3424 | 60 |
AB197664.1 | - | 4 - 5 | 3591 | 60 | |
AK024475.1 | - | 6 - 7 | 1123 | 60 | |
AK304723.1 | - | 4 - 5 | 579 | 60 | |
AK308298.1 | - | 6 - 7 | 1097 | 60 | |
AL117435.1 | - | 1 - 2 | 60 | 60 | |
BC054486.1 | - | 4 - 5 | 1102 | 60 | |
BC063501.1 | - | 6 - 7 | 1099 | 60 | |
BC082974.1 | - | 7 - 8 | 1196 | 60 | |
BC115390.1 | - | 4 - 5 | 1205 | 60 |