Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IKZF2
Entrez Gene ID
22807
Gene Name
IKAROS family zinc finger 2
Gene Aliases
ANF1A2, HELIOS, ZNF1A2, ZNFN1A2
Chromosome Location
Chr.2: 212999684 - 213152753 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079526.1 NP_001072994.1 1 - 2 203 91
XM_011510802.2 XP_011509104.1 1 - 2 272 91
XM_011510807.2 XP_011509109.1 1 - 2 271 91
GenBank mRNA BC028936.1 - 1 - 2 203 91

View Full Product Details