Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNS1
Entrez Gene ID
7145
Gene Name
tensin 1
Gene Aliases
MST091, MST122, MST127, MSTP091, MSTP122, MSTP127, MXRA6, PPP1R155, TNS
Chromosome Location
Chr.2: 217799789 - 217978853 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308022.1 NP_001294951.1 21 - 22 3415 71
NM_001308023.1 NP_001294952.1 20 - 21 3391 71
NM_022648.5 NP_072174.3 23 - 24 3663 71
XM_011511712.1 XP_011510014.1 21 - 22 3646 71
XM_011511713.1 XP_011510015.1 21 - 22 3631 71
XM_011511715.1 XP_011510017.1 20 - 21 3607 71
XM_011511723.2 XP_011510025.1 21 - 22 3607 71
XM_011511728.1 XP_011510030.1 5 - 6 4699 71
XM_017004801.1 XP_016860290.1 22 - 23 3802 71
XM_017004802.1 XP_016860291.1 21 - 22 3754 71
XM_017004803.1 XP_016860292.1 21 - 22 3715 71
XM_017004804.1 XP_016860293.1 22 - 23 3670 71
XM_017004805.1 XP_016860294.1 21 - 22 3586 71
XM_017004806.1 XP_016860295.1 20 - 21 3559 71
XM_017004807.1 XP_016860296.1 19 - 20 3520 71
XM_017004808.1 XP_016860297.1 18 - 19 3295 71
XM_017004809.1 XP_016860298.1 26 - 27 4143 71
XM_017004810.1 XP_016860299.1 26 - 27 4026 71
XM_017004811.1 XP_016860300.1 24 - 25 3889 71
XM_017004812.1 XP_016860301.1 23 - 24 3839 71
XM_017004813.1 XP_016860302.1 9 - 10 2751 71
XM_017004814.1 XP_016860303.1 9 - 10 2539 71
XM_017004815.1 XP_016860304.1 9 - 10 2605 71
XM_017004816.1 XP_016860305.1 2 - 3 2914 71
XM_017004817.1 XP_016860306.1 7 - 8 4894 71
GenBank mRNA AB209238.1 - 5 - 6 679 71
AK300345.1 - 21 - 22 3597 71
BC116187.1 - 20 - 21 3388 71
BC116188.1 - 19 - 20 3361 71
BC126910.1 - 19 - 20 3364 71
BC140942.1 - 23 - 24 3595 71

View Full Product Details