Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNS1
Entrez Gene ID
7145
Gene Name
tensin 1
Gene Aliases
MST091, MST122, MST127, MSTP091, MSTP122, MSTP127, MXRA6, PPP1R155, TNS
Chromosome Location
Chr.2: 217799789 - 217978853 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308022.1 NP_001294951.1 25 - 26 4788 66
NM_001308023.1 NP_001294952.1 24 - 25 4767 66
NM_022648.5 NP_072174.3 27 - 28 5039 66
XM_011511712.1 XP_011510014.1 25 - 26 5022 66
XM_011511713.1 XP_011510015.1 25 - 26 5007 66
XM_011511715.1 XP_011510017.1 24 - 25 4983 66
XM_011511723.2 XP_011510025.1 25 - 26 4983 66
XM_011511728.1 XP_011510030.1 9 - 10 6075 66
XM_017004801.1 XP_016860290.1 26 - 27 5175 66
XM_017004802.1 XP_016860291.1 25 - 26 5130 66
XM_017004803.1 XP_016860292.1 25 - 26 5091 66
XM_017004804.1 XP_016860293.1 26 - 27 5046 66
XM_017004805.1 XP_016860294.1 25 - 26 4962 66
XM_017004806.1 XP_016860295.1 24 - 25 4935 66
XM_017004807.1 XP_016860296.1 23 - 24 4896 66
XM_017004808.1 XP_016860297.1 22 - 23 4671 66
XM_017004809.1 XP_016860298.1 30 - 31 5519 66
XM_017004810.1 XP_016860299.1 30 - 31 5402 66
XM_017004811.1 XP_016860300.1 28 - 29 5265 66
XM_017004812.1 XP_016860301.1 27 - 28 5215 66
XM_017004813.1 XP_016860302.1 13 - 14 4127 66
XM_017004814.1 XP_016860303.1 13 - 14 3915 66
XM_017004815.1 XP_016860304.1 13 - 14 3981 66
XM_017004816.1 XP_016860305.1 6 - 7 4290 66
XM_017004817.1 XP_016860306.1 11 - 12 6270 66
GenBank mRNA AB209238.1 - 9 - 10 2055 66
AL117537.1 - 3 - 4 247 66
BC014042.2 - 4 - 5 751 66
BC116187.1 - 24 - 25 4761 66
BC116188.1 - 23 - 24 4737 66
BC126910.1 - 23 - 24 4740 66
BC140942.1 - 27 - 28 4971 66

View Full Product Details