The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324051.1 | NP_001310980.1 | 13 - 14 | 1939 | 99 |
NM_001324052.1 | NP_001310981.1 | 15 - 16 | 2129 | 99 | |
NM_001324053.1 | NP_001310982.1 | 15 - 16 | 2288 | 99 | |
NM_001324054.1 | NP_001310983.1 | 13 - 14 | 1919 | 99 | |
NM_173206.3 | NP_775298.1 | 12 - 13 | 1830 | 99 | |
NR_136685.1 | 11 - 12 | 1355 | 99 | ||
XM_006722573.2 | XP_006722636.1 | 13 - 14 | 1813 | 99 | |
XM_017026071.1 | XP_016881560.1 | 13 - 14 | 1744 | 99 | |
XM_017026072.1 | XP_016881561.1 | 12 - 13 | 1683 | 99 | |
GenBank mRNA | AF077953.1 | - | 12 - 13 | 1643 | 99 |
AK294801.1 | - | 11 - 12 | 1328 | 99 | |
AW016227.1 | - | 1 - 2 | 99 | ||
BC015190.1 | - | 12 - 13 | 1801 | 99 |