The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014938.5 | NP_055753.3 | 8 - 9 | 1217 | 85 |
XM_006719290.3 | XP_006719353.1 | 8 - 9 | 1201 | 85 | |
XM_006719291.3 | XP_006719354.1 | 8 - 9 | 1201 | 85 | |
XM_006719292.3 | XP_006719355.1 | 8 - 9 | 1201 | 85 | |
XM_006719293.3 | XP_006719356.1 | 8 - 9 | 1201 | 85 | |
GenBank mRNA | AF312918.1 | - | 8 - 9 | 1091 | 85 |
BC028309.1 | - | 6 - 7 | 566 | 85 | |
BC069272.1 | - | 3 - 4 | 354 | 85 | |
BC151841.1 | - | 8 - 9 | 1199 | 85 |