The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242359.1 | NP_001229288.1 | 10 - 11 | 2254 | 75 |
NM_014836.4 | NP_055651.1 | 9 - 10 | 2182 | 75 | |
NR_024554.2 | 10 - 11 | 2056 | 75 | ||
NR_024555.1 | 10 - 11 | 2020 | 75 | ||
XM_006718084.1 | XP_006718147.1 | 9 - 10 | 1990 | 75 | |
XM_006718085.1 | XP_006718148.1 | 9 - 10 | 2017 | 75 | |
XM_011540424.1 | XP_011538726.1 | 10 - 11 | 2048 | 75 | |
XM_017017000.1 | XP_016872489.1 | 9 - 10 | 2237 | 75 | |
XM_017017001.1 | XP_016872490.1 | 9 - 10 | 2233 | 75 | |
XM_017017003.1 | XP_016872492.1 | 11 - 12 | 2123 | 75 | |
XM_017017004.1 | XP_016872493.1 | 11 - 12 | 2125 | 75 | |
XM_017017005.1 | XP_016872494.1 | 10 - 11 | 2030 | 75 | |
XM_017017006.1 | XP_016872495.1 | 10 - 11 | 2079 | 75 | |
XM_017017007.1 | XP_016872496.1 | 8 - 9 | 2129 | 75 | |
XM_017017008.1 | XP_016872497.1 | 10 - 11 | 2290 | 75 | |
GenBank mRNA | AB018283.3 | - | 9 - 10 | 2073 | 75 |
AK075129.1 | - | 10 - 11 | 2020 | 75 | |
AK307630.1 | - | 8 - 9 | 1752 | 75 | |
BC032848.1 | - | 10 - 11 | 2231 | 75 | |
BC041791.1 | - | 9 - 10 | 1973 | 75 |