The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321962.1 | NP_001308891.1 | 6 - 7 | 845 | 123 |
NM_001321963.1 | NP_001308892.1 | 8 - 9 | 1516 | 123 | |
NM_001321964.1 | NP_001308893.1 | 8 - 9 | 1841 | 123 | |
NM_001321965.1 | NP_001308894.1 | 9 - 10 | 2154 | 123 | |
NM_012472.5 | NP_036604.2 | 8 - 9 | 1091 | 123 | |
NR_073525.2 | 8 - 9 | 1091 | 123 | ||
NR_135905.1 | 8 - 9 | 1180 | 123 | ||
NR_135906.1 | 5 - 6 | 621 | 123 | ||
NR_135907.1 | 7 - 8 | 867 | 123 | ||
NR_135909.1 | 7 - 8 | 985 | 123 | ||
NR_135910.1 | 7 - 8 | 1292 | 123 | ||
NR_135911.1 | 5 - 6 | 1371 | 123 | ||
NR_135912.1 | 8 - 9 | 1930 | 123 | ||
NR_135913.1 | 7 - 8 | 1617 | 123 | ||
XM_006716538.3 | XP_006716601.2 | 8 - 9 | 1055 | 123 | |
XM_017013296.1 | XP_016868785.1 | 7 - 8 | 1724 | 123 | |
XM_017013297.1 | XP_016868786.1 | 8 - 9 | 1209 | 123 | |
XM_017013298.1 | XP_016868787.1 | 10 - 11 | 1136 | 123 | |
GenBank mRNA | AK313107.1 | - | 8 - 9 | 1066 | 123 |
BC027589.1 | - | 8 - 9 | 1037 | 123 | |
BC047286.1 | - | 8 - 9 | 1041 | 123 |