The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020120.3 | NP_064505.1 | 19 - 20 | 2312 | 95 |
NR_027671.2 | 19 - 20 | 2465 | 95 | ||
XM_006712634.3 | XP_006712697.1 | 19 - 20 | 2286 | 95 | |
XM_006712635.3 | XP_006712698.1 | 19 - 20 | 2280 | 95 | |
XM_006712636.3 | XP_006712699.1 | 19 - 20 | 2275 | 95 | |
XM_017004507.1 | XP_016859996.1 | 20 - 21 | 2371 | 95 | |
XM_017004508.1 | XP_016859997.1 | 20 - 21 | 2326 | 95 | |
XM_017004509.1 | XP_016859998.1 | 5 - 6 | 617 | 95 | |
GenBank mRNA | AF227905.1 | - | 19 - 20 | 2172 | 95 |
AK074251.1 | - | 3 - 4 | 336 | 95 | |
AK293066.1 | - | 19 - 20 | 2465 | 95 | |
AK309101.1 | - | 17 - 18 | 2037 | 95 | |
BC041098.1 | - | 19 - 20 | 2462 | 95 |