Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UGGT1
Entrez Gene ID
56886
Gene Name
UDP-glucose glycoprotein glucosyltransferase 1
Gene Aliases
HUGT1, UGCGL1, UGT1
Chromosome Location
Chr.2: 128091180 - 128195677 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020120.3 NP_064505.1 29 - 30 3473 108
NR_027671.2 29 - 30 3626 108
XM_006712634.3 XP_006712697.1 29 - 30 3447 108
XM_006712635.3 XP_006712698.1 28 - 29 3417 108
XM_006712636.3 XP_006712699.1 29 - 30 3436 108
XM_017004507.1 XP_016859996.1 30 - 31 3532 108
XM_017004508.1 XP_016859997.1 30 - 31 3487 108
XM_017004509.1 XP_016859998.1 15 - 16 1778 108
XM_017004510.1 XP_016859999.1 11 - 12 2157 108
GenBank mRNA AF227905.1 - 29 - 30 3333 108
AK023671.1 - 10 - 11 1169 108
AK074251.1 - 13 - 14 1497 108
AK074376.1 - 3 - 4 468 108
AK293066.1 - 29 - 30 3626 108
BC041098.1 - 29 - 30 3623 108

View Full Product Details