Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D2
Entrez Gene ID
55357
Gene Name
TBC1 domain family member 2
Gene Aliases
PARIS-1, PARIS1, TBC1D2A
Chromosome Location
Chr.9: 98198998 - 98255721 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267571.1 NP_001254500.1 2 - 3 688 83
NM_018421.3 NP_060891.3 2 - 3 688 83
XM_011518844.2 XP_011517146.1 2 - 3 688 83
GenBank mRNA AB209721.1 - 2 - 3 488 83
AB449882.1 - 2 - 3 508 83
AF318370.1 - 2 - 3 547 83
AK131290.1 - 2 - 3 566 83
AK298575.1 - 2 - 3 359 83
AY026527.1 - 2 - 3 600 83

View Full Product Details