The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267571.1 | NP_001254500.1 | 3 - 4 | 826 | 74 |
NM_018421.3 | NP_060891.3 | 3 - 4 | 826 | 74 | |
NR_051978.1 | 2 - 3 | 684 | 74 | ||
XM_011518844.2 | XP_011517146.1 | 3 - 4 | 826 | 74 | |
XM_017014899.1 | XP_016870388.1 | 1 - 2 | 128 | 74 | |
GenBank mRNA | AB209721.1 | - | 3 - 4 | 626 | 74 |
AB449882.1 | - | 3 - 4 | 646 | 74 | |
AF318370.1 | - | 3 - 4 | 685 | 74 | |
AK026105.1 | - | 2 - 3 | 174 | 74 | |
AK124772.1 | - | 2 - 3 | 1375 | 74 | |
AK131290.1 | - | 3 - 4 | 704 | 74 | |
AK298575.1 | - | 4 - 5 | 476 | 74 | |
AY026527.1 | - | 3 - 4 | 738 | 74 |