The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031701.2 | NP_001026871.1 | 3 - 4 | 513 | 127 |
XM_017019454.1 | XP_016874943.1 | 3 - 4 | 554 | 127 | |
GenBank mRNA | AB032773.1 | - | 3 - 4 | 323 | 127 |
BC052293.1 | - | 3 - 4 | 513 | 127 |