Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NT5DC3
Entrez Gene ID
51559
Gene Name
5'-nucleotidase domain containing 3
Gene Aliases
TU12B1-TY, TU12B1TY
Chromosome Location
Chr.12: 103746315 - 103841238 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031701.2 NP_001026871.1 10 - 11 1146 75
XM_011538476.2 XP_011536778.1 7 - 8 903 75
GenBank mRNA AB032773.1 - 10 - 11 956 75
BC052293.1 - 10 - 11 1146 75

View Full Product Details