The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000943.4 | NP_000934.1 | 3 - 4 | 428 | 111 |
GenBank mRNA | AK290321.1 | - | 3 - 4 | 386 | 111 |
BC002678.2 | - | 3 - 4 | 401 | 111 | |
BG721394.1 | - | 3 - 4 | 428 | 111 | |
S71018.1 | - | 3 - 4 | 487 | 111 |