The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014433.2 | NP_001014433.1 | 3 - 4 | 470 | 66 |
NM_001014837.1 | NP_001014837.1 | 3 - 4 | 719 | 66 | |
NM_001014838.1 | NP_001014838.1 | 3 - 4 | 688 | 66 | |
NM_001014840.1 | NP_001014840.1 | 3 - 4 | 598 | 66 | |
NM_015921.2 | NP_057005.1 | 2 - 3 | 861 | 66 | |
GenBank mRNA | AF106943.1 | - | 2 - 3 | 498 | 66 |
AF230924.1 | - | 2 - 3 | 284 | 66 | |
BC005890.2 | - | 3 - 4 | 295 | 66 | |
BC107751.1 | - | 2 - 3 | 498 | 66 | |
BU540844.1 | - | - | 476 | 66 | |
CB106834.1 | - | 3 - 4 | 424 | 66 | |
CN363642.1 | - | 3 - 4 | 500 | 66 |