The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289968.1 | NP_001276897.1 | 11 - 12 | 1373 | 104 |
NM_001289969.1 | NP_001276898.1 | 11 - 12 | 1438 | 104 | |
NM_021132.3 | NP_066955.1 | 10 - 11 | 1319 | 104 | |
XM_005269944.1 | XP_005270001.1 | 10 - 11 | 1319 | 104 | |
GenBank mRNA | AK300701.1 | - | 2 - 3 | 115 | 104 |
BC035464.1 | - | 10 - 11 | 1277 | 104 | |
M29550.1 | - | 11 - 12 | 1345 | 104 | |
M29551.1 | - | 10 - 11 | 1300 | 104 |