The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320540.1 | NP_001307469.1 | 9 - 10 | 790 | 74 |
NM_001320541.1 | NP_001307470.1 | 9 - 10 | 831 | 74 | |
NM_001320542.1 | NP_001307471.1 | 9 - 10 | 816 | 74 | |
NM_001320543.1 | NP_001307472.1 | 8 - 9 | 747 | 74 | |
NM_025187.4 | NP_079463.2 | 8 - 9 | 846 | 74 | |
XM_006721284.3 | XP_006721347.1 | 8 - 9 | 1105 | 74 | |
XM_011523350.2 | XP_011521652.1 | 7 - 8 | 1245 | 74 | |
XM_017023729.1 | XP_016879218.1 | 8 - 9 | 1116 | 74 | |
GenBank mRNA | AF176088.1 | - | 9 - 10 | 746 | 74 |
AL540304.3 | - | 9 - 10 | 725 | 74 | |
BC004556.2 | - | 8 - 9 | 826 | 74 | |
CB997191.1 | - | 8 - 9 | 705 | 74 |