The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052882.1 | NP_443114.1 | 3 - 4 | 529 | 73 |
GenBank mRNA | AF365931.1 | - | 3 - 4 | 529 | 73 |
BC069071.1 | - | 1 - 2 | 144 | 73 | |
BC114446.1 | - | 3 - 4 | 328 | 73 | |
BC114503.1 | - | 3 - 4 | 328 | 73 |