The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015657.3 | NP_056472.2 | 26 - 27 | 4580 | 81 |
NM_173076.2 | NP_775099.2 | 34 - 35 | 5595 | 81 | |
NR_103740.1 | 36 - 37 | 5675 | 81 | ||
XM_011510951.2 | XP_011509253.1 | 34 - 35 | 5767 | 81 | |
GenBank mRNA | AF418105.1 | - | 29 - 30 | 4717 | 81 |
AK123578.1 | - | 1 - 2 | 134 | 81 | |
AY033486.1 | - | 26 - 27 | 4580 | 81 | |
AY219711.1 | - | 34 - 35 | 5595 | 81 |