Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABCA12
Entrez Gene ID
26154
Gene Name
ATP binding cassette subfamily A member 12
Gene Aliases
ARCI4A, ARCI4B, ICR2B, LI2
Chromosome Location
Chr.2: 214931542 - 215138591 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015657.3 NP_056472.2 30 - 31 4979 116
NM_173076.2 NP_775099.2 38 - 39 5994 116
NR_103740.1 40 - 41 6074 116
XM_011510951.2 XP_011509253.1 38 - 39 6166 116
GenBank mRNA AF418105.1 - 33 - 34 5116 116
AY033486.1 - 30 - 31 4979 116
AY219711.1 - 38 - 39 5994 116

View Full Product Details