The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015657.3 | NP_056472.2 | 43 - 44 | 6750 | 79 |
NM_173076.2 | NP_775099.2 | 51 - 52 | 7765 | 79 | |
NR_103740.1 | 53 - 54 | 7845 | 79 | ||
XM_011510951.2 | XP_011509253.1 | 51 - 52 | 7937 | 79 | |
GenBank mRNA | AF418105.1 | - | 46 - 47 | 6887 | 79 |
AK096597.1 | - | 7 - 8 | 890 | 79 | |
AK123578.1 | - | 18 - 19 | 2305 | 79 | |
AL080207.1 | - | 4 - 5 | 349 | 79 | |
AY033486.1 | - | 43 - 44 | 6750 | 79 | |
AY219711.1 | - | 51 - 52 | 7765 | 79 |