The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270523.1 | NP_001257452.1 | 3 - 4 | 384 | 69 |
NM_001270524.1 | NP_001257453.1 | 2 - 3 | 537 | 69 | |
NM_172337.2 | NP_758840.1 | 1 - 2 | 307 | 69 | |
GenBank mRNA | AB593057.1 | - | 1 - 2 | 77 | 69 |
AF093138.1 | - | 1 - 2 | 100 | 69 | |
AK314271.1 | - | 2 - 3 | 440 | 69 | |
BE781530.1 | - | 1 - 2 | 271 | 69 | |
CV812961.1 | - | 3 - 4 | 361 | 69 |