Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MICAL1
Entrez Gene ID
64780
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 1
Gene Aliases
MICAL, MICAL-1, NICAL
Chromosome Location
Chr.6: 109444062 - 109465968 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159291.1 NP_001152763.1 20 - 21 2842 76
NM_001286613.1 NP_001273542.1 21 - 22 3136 76
NM_022765.3 NP_073602.3 21 - 22 3100 76
GenBank mRNA AB048948.1 - 21 - 22 3100 76
AK021999.1 - 20 - 21 2843 76
AK024500.1 - 5 - 6 3865 76
AK160384.1 - 18 - 19 3787 76
AK225049.1 - 21 - 22 2857 76
AK296284.1 - 21 - 22 3139 76
AL122098.1 - 13 - 14 1526 76
BC009972.2 - 20 - 21 2597 76
BC042144.1 - 21 - 22 2996 76
BC052983.1 - 20 - 21 2813 76

View Full Product Details