Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BRPF3
Entrez Gene ID
27154
Gene Name
bromodomain and PHD finger containing 3
Gene Aliases
-
Chromosome Location
Chr.6: 36196773 - 36232790 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011514489.1 XP_011512791.1 1 - 2 103 127

View Full Product Details