The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009590.2 | NP_033720.2 | 2 - 3 | 1914 | 84 |
GenBank mRNA | AF081363.1 | - | 2 - 3 | 1905 | 84 |
BC142641.1 | - | 2 - 3 | 1906 | 84 |