The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023935.2 | NP_076424.1 | 1 - 2 | 160 | 85 |
GenBank mRNA | AK310809.1 | - | 1 - 2 | 143 | 85 |
BC000643.1 | - | 1 - 2 | 112 | 85 | |
BC007957.1 | - | 1 - 2 | 118 | 85 | |
BC011851.1 | - | 1 - 2 | 113 | 85 | |
HY128886.1 | - | 1 - 2 | 85 |