Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANO7
Entrez Gene ID
50636
Gene Name
anoctamin 7
Gene Aliases
D-TMPP, DTMPP, IPCA-5, IPCA5, NGEP, PCANAP5, PCANAP5L, TMEM16G
Chromosome Location
Chr.2: 241188509 - 241239602 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC028162.1 - 2 - 2 731 83
BC038852.1 - 2 - 2 723 83
BC047903.1 - 2 - 2 748 83

View Full Product Details