The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168551.1 | NP_001162023.1 | 5 - 6 | 762 | 113 |
NM_018087.4 | NP_060557.3 | 5 - 6 | 882 | 113 | |
NR_033142.1 | 4 - 5 | 761 | 113 | ||
XM_011541766.2 | XP_011540068.1 | 5 - 6 | 724 | 113 | |
GenBank mRNA | AK001269.1 | - | 5 - 6 | 765 | 113 |
AK295000.1 | - | 5 - 6 | 546 | 113 | |
AK298909.1 | - | 4 - 5 | 505 | 113 | |
AK302910.1 | - | 4 - 5 | 476 | 113 | |
AL354612.1 | - | 5 - 6 | 765 | 113 | |
AL354613.1 | - | 5 - 6 | 762 | 113 | |
BC003082.2 | - | 5 - 6 | 832 | 113 | |
BX648031.1 | - | 4 - 5 | 492 | 113 | |
DQ141696.1 | - | 5 - 6 | 591 | 113 |